Learn More
Waters Corp Waters Sample Loop, 5 μL, For Rheodyne Injectors
Catalog No. 50793788 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This sample loop is designed for use with 7010 and 7125 Rheodyne injectors. It provides precise 5 μL sample delivery for various applications.
  • Used on 7010 and 7125 Rheodyne injectors
  • Provides 5 μL sample volume
  • Suitable for HPLC systems
  • Replaced as required for optimal performance
Catalog No. 50-793-788 Supplier Waters Corp Supplier No. WAT096223
This sample loop is designed for use with 7010 and 7125 Rheodyne injectors. It provides precise 5 μL sample delivery for various applications.
  • Used on 7010 and 7125 Rheodyne injectors
  • Provides 5 μL sample volume
  • Suitable for HPLC systems
  • Replaced as required for optimal performance