Learn More
Trajan Scientific VICI Valco Sample Loop Valco 5l Stainless Steel (VAL.SL5CW)
Catalog No. 50635029
Encompass
VICI Valco 1/16" Stainless Steel Loops Includes Two Stainless Nuts and Two Stainless ferrµles. VICI Valco Part Number: SL5CW (NEW LPP PN: VAL.SL5CW). Thermo Part Number: CH-952453. Made in the USA.
Catalog No. 50-635-029 Supplier Trajan Scientific Supplier No. PAL.SL5CW
VICI Valco 1/16" Stainless Steel Loops Includes Two Stainless Nuts and Two Stainless ferrµles. VICI Valco Part Number: SL5CW (NEW LPP PN: VAL.SL5CW). Thermo Part Number: CH-952453. Made in the USA.