Learn More
Waters Corp Waters Sample Loop, Ext. Hypo Tip, 5 μL
Catalog No. NC2195330 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This sample loop is designed for precise fluid handling in laboratory environments. Featuring an extended hypo tip, it ensures accurate and reliable sample introduction.
  • Features an extended hypo tip.
  • Provides a 5 μL sample volume.
  • Compatible with ACQUITY I-Class PLUS Sample Manager-FL system.
  • Compatible with ACQUITY UPC2 SM-FL system.
This item is not returnable. View return policy
Catalog No. NC2195330 Supplier Waters Corp Supplier No. 430002936
This sample loop is designed for precise fluid handling in laboratory environments. Featuring an extended hypo tip, it ensures accurate and reliable sample introduction.
  • Features an extended hypo tip.
  • Provides a 5 μL sample volume.
  • Compatible with ACQUITY I-Class PLUS Sample Manager-FL system.
  • Compatible with ACQUITY UPC2 SM-FL system.