Learn More
Thermo Scientific SST Sample Loops for Split-Loop Autosamplers, 25 μL
Catalog No. 501348250 Shop All Thermo Scientific Products
Encompass
Sterile, single-use 25 microliter, stainless steel (SST) sample loops incorporate Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.
Catalog No. 50-134-8250 Supplier Thermo Scientific Supplier No. 6820.2452
Sterile, single-use 25 microliter, stainless steel (SST) sample loops incorporate Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.