Learn More
Agilent Technologies Sample Loop Assembly, Stainless Steel, 100 μL, 1 ea
Catalog No. NC3356905
Encompass
This stainless steel sample loop assembly has a volume of 100 μL and is designed for specific chromatography applications.
  • Volume: 100 μL
  • Material: Stainless steel
  • Compatibility: Engineered for use with G7167A/B Multisampler systems in a dual needle, left-side configuration
Catalog No. NC3356905 Supplier Agilent Technologies Supplier No. G426760501
This stainless steel sample loop assembly has a volume of 100 μL and is designed for specific chromatography applications.
  • Volume: 100 μL
  • Material: Stainless steel
  • Compatibility: Engineered for use with G7167A/B Multisampler systems in a dual needle, left-side configuration