Learn More
Trajan Scientific VICI Valco Sample Loop, 2-L, Cheminert, Stainless Steel, Loop With 1/16"" Tube Ends Includes Two Stainless Steel Nuts and Ferrules
Catalog No. 501090212
Encompass
VICI Valco Sample Loop, 2-¿L, Cheminert, Stainless Steel Loop With 1/16" Tube Ends Includes Two SS Nuts and Ferrules. Made in the USA.
Catalog No. 50-109-0212 Supplier Trajan Scientific Supplier No. CSL2
VICI Valco Sample Loop, 2-¿L, Cheminert, Stainless Steel Loop With 1/16" Tube Ends Includes Two SS Nuts and Ferrules. Made in the USA.