Learn More
Perkin Elmer US LLC Sample Loop for 10-Port Valve, Volume 5 mL, Fitting Size: 1/16 in
Catalog No. 502112853
Encompass_Preferred
External Volume sample size is determined by a sample loop outside the valve. 1/8 in. fitting loop for valves with 0. 030 in. standard port diameter. 1/16 in. fitting loop for valves with 0. 016 in. standard port diameter.
Catalog No. 50-211-2853 Supplier Perkin Elmer US LLC Supplier No. N9302958
External Volume sample size is determined by a sample loop outside the valve. 1/8 in. fitting loop for valves with 0. 030 in. standard port diameter. 1/16 in. fitting loop for valves with 0. 016 in. standard port diameter.