Learn More
Perkin Elmer US LLC Sample Loop for 10-Port Valve, Volume 100µL, Fitting Size: 1/16 in
Catalog No. 502112761
Encompass_Preferred
1/8 in. fitting loop for valves with 0.030 in. standard port diameter. 1/16 in. fitting loop for valves with 0.016 in. standard port diameter.
Catalog No. 50-211-2761 Supplier Perkin Elmer US LLC Supplier No. N9302857
1/8 in. fitting loop for valves with 0.030 in. standard port diameter. 1/16 in. fitting loop for valves with 0.016 in. standard port diameter.