Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perkin Elmer US LLC 25 mL LVA PEEK Sample Loop

Catalog No. 502016568
Encompass_Preferred

Sample Loop, Peek, 25ml Aquatek Lva

Catalog No. 50-201-6568 Supplier Perkin Elmer US LLC Supplier No. N6353111
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sample Loop, Peek, 25ml Aquatek Lva

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.