Learn More
Perkin Elmer US LLC Sample Needle with Fittings for Syringe Kits
Catalog No. 502112579
Encompass_Preferred
PerkinElmer syringe kits for high pressure applications are manufactured for precise liquid delivery. All of these glass syringes come with precision stainless steel plungers. They are used for sampling and flushing and are available in a large variety of sizes.
Catalog No. 50-211-2579 Supplier Perkin Elmer US LLC Supplier No. N2936009
PerkinElmer syringe kits for high pressure applications are manufactured for precise liquid delivery. All of these glass syringes come with precision stainless steel plungers. They are used for sampling and flushing and are available in a large variety of sizes.