Learn More
Cole-Parmer Wildco 425-D52 D-frame Multifilment 500 um (EPA) Net
Supplier:  Cole-Parmer 0549132
Encompass_Preferred
Stainless steel net ring and removable hardwood handle will provide years of use. Canvas shroud on bottom of net protects it from snags while still allowing clear view of contents.
Catalog No. NC2109592