Learn More
AirSea USA Sample Pack. UN Fiberboard box for various inners, 4G/X2/S, Int Dims: 2 3/4"" x 2 3/4"" x 6 7/8""
Catalog No. 501534828
Encompass_Preferred
Sample Pack - UN 4G Boxes. UN Mark: 4G/X2/S. External Dimensions: 105mm x 105mm x 220mm. Our sample pack 4G boxes are compliant withDOT IATA and ICAO dangerous goods regulations meaning that they are suitable for shipments of dangerous goods via air, as well as meeting ADR requirements for road and IMDG stndards for transportation via sea. This 4G box is approved for shipping both solid and liquid dangerous goods.
Catalog No. 50-153-4828 Supplier AirSea USA Supplier No. 474
Sample Pack - UN 4G Boxes. UN Mark: 4G/X2/S. External Dimensions: 105mm x 105mm x 220mm. Our sample pack 4G boxes are compliant withDOT IATA and ICAO dangerous goods regulations meaning that they are suitable for shipments of dangerous goods via air, as well as meeting ADR requirements for road and IMDG stndards for transportation via sea. This 4G box is approved for shipping both solid and liquid dangerous goods.