Learn More
Perkin Elmer US LLC Sample Pan - Platinum/Rhodium, 5.0 mm, Qty. 1
Catalog No. 502104340
Encompass_Preferred
This Sample Pan is made of a combination of platinum and rhodium to form a corrosion-resistant, chemically-inert sample pan. This pan is 5.0 mm high and is compatible with the TGA 4000 and STA 6000/8000 analyzers. This pan can be purchased as a spare set using this part number. Qty. 1
Catalog No. 50-210-4340 Supplier Perkin Elmer US LLC Supplier No. N5201024
This Sample Pan is made of a combination of platinum and rhodium to form a corrosion-resistant, chemically-inert sample pan. This pan is 5.0 mm high and is compatible with the TGA 4000 and STA 6000/8000 analyzers. This pan can be purchased as a spare set using this part number. Qty. 1