Learn More
Perkin Elmer US LLC Sample Pan - Platinum/Rhodium, 5.0 mm, Qty. 5
Catalog No. 502104342
Encompass_Preferred
This Sample Pan is made of a combination of platinum and rhodium to form a corrosion-resistant, chemically-inert sample pan. This pan is 5.0 mm high and is compatible with the TGA 4000 and STA 6000/8000 analyzers. This pan can be purchased as a spare set using this part number. Qty. 5
Catalog No. 50-210-4342 Supplier Perkin Elmer US LLC Supplier No. N5201022
This Sample Pan is made of a combination of platinum and rhodium to form a corrosion-resistant, chemically-inert sample pan. This pan is 5.0 mm high and is compatible with the TGA 4000 and STA 6000/8000 analyzers. This pan can be purchased as a spare set using this part number. Qty. 5