Learn More
Research Products International Corp Micro-Tube Sample Pestles for 1.5ml Tubes, RNase and DNase Free, 100 per Package
SDP
Catalog No. 50476500
Encompass
Ideal for manually homogenizing cells and resuspending proteins or DNA pellets in buffers. Polypropylene and autoclavable.
Pestle only for 1.5ml tubes.
Catalog No. 50-476-500 Supplier Research Products International Corp Supplier No. 199228
Ideal for manually homogenizing cells and resuspending proteins or DNA pellets in buffers. Polypropylene and autoclavable.
Pestle only for 1.5ml tubes.