Learn More
AB Sciex LLC SAMPLE PLATE PACK
Catalog No. NC2157555
Encompass
BioPhase 8800 Sample plate pack includes 20 sample plates. Plates include 96 wells and are designed to ANSI standard configuration for compatibility with automated 96-well plate handlers.Plate design enables use of 8-capillary BioPhase cartridges.
This item is not returnable. View return policy
Catalog No. NC2157555 Supplier AB Sciex LLC Supplier No. 5080313
BioPhase 8800 Sample plate pack includes 20 sample plates. Plates include 96 wells and are designed to ANSI standard configuration for compatibility with automated 96-well plate handlers.Plate design enables use of 8-capillary BioPhase cartridges.