Learn More
Protifi LLC Universal proteomics sample preparation kits, minis (100 µg - 300 µg) with solutions
SDP
Supplier:  Protifi LLC K02MINI80
Encompass
S-Trap mini sample processing in a full kit format for samples from 100 µg - 300 µg. The kit includes S-Trap minis, extra tubes, and lysis, wash and digestion buffers. The user must supply HPLC or MS grade MeOH and their choice of protease.
Catalog No. NC3050531