Learn More
Anton Paar Sample presser for solid samples for Abbemat 300/350/500/550
Catalog No. NC1818545
Encompass_Preferred
The sample presser is used for solid samples, e.g. plastic films. It ensures a coplanar orientation of the sample on the prism, even if the sample is flexible.Suitable for use with Anton Paar refractometersAbbemat 300 item NC1818534,Abbemat 350 item NC1818535,Abbemat 500 item NC1818537,Abbemat 550 item NC1818539
Catalog No. NC1818545 Supplier Anton Paar Supplier No. 110284
The sample presser is used for solid samples, e.g. plastic films. It ensures a coplanar orientation of the sample on the prism, even if the sample is flexible.Suitable for use with Anton Paar refractometersAbbemat 300 item NC1818534,Abbemat 350 item NC1818535,Abbemat 500 item NC1818537,Abbemat 550 item NC1818539