Learn More
Perkin Elmer US LLC Sample Probe Cleaning Wire, Pkg. 1
Catalog No. 502112516
Encompass_Preferred
Note that the outer diameter of the cleaning wires is 0.4 mm, because of this it is strongly recommended to be used only with sample probes with inner diameters of 0.6 mm or larger.
Catalog No. 50-211-2516 Supplier Perkin Elmer US LLC Supplier No. B0505962
Note that the outer diameter of the cleaning wires is 0.4 mm, because of this it is strongly recommended to be used only with sample probes with inner diameters of 0.6 mm or larger.