Learn More
Perkin Elmer US LLC FIAS Sampling Probe Assembly for PerkinElmer AS-90/90A/90 Plus/91/93 Plus Autosampler, 0.6mm Inner Diameter, Replaces P/N B0196966
Catalog No. 509042749
Encompass_Preferred
Includes Stainless Steel Sampling Probe and Screw Fitting (B3000152) and FIAS Capillary Tubing Assembly (B3000158 – Replaces B0196966).
Catalog No. 50-904-2749 Supplier Perkin Elmer US LLC Supplier No. B3000160
Includes Stainless Steel Sampling Probe and Screw Fitting (B3000152) and FIAS Capillary Tubing Assembly (B3000158 – Replaces B0196966).