Learn More
Thermo Scientific Sample Probe for ICP
Supplier:  Thermo Scientific 842312051501
Encompass
Sample probe introduces liquid into ICP-MS for elemental analysis by drawing sample through capillary tubing with peristaltic pump, inserting probe tip into plasma torch where sample desolvated, atomized, ionized then analyzed by mass spectrometer to determine composition, made of inert materials.
Catalog No. NC1273592