Learn More
Thermo Scientific Vanquish™ Charger and Split Sampler Sample Racks
Catalog No. NC1368072 Shop All Thermo Scientific Products
Encompass
Sample rack, 54 pos, 12 mm OD vials, Thermo Scientific™ Vanquish™ Charger and Split Sampler Sample racks are for use in the Vanquish Split Sampler modules, and fit into the 9-shelf of the Charger module, which is an intelligent robotic that automates sample loading into the Thermo Scientific Vanquish UHPLC systems for extra-long unattended operation.
This item is not returnable. View return policy
Catalog No. NC1368072 Supplier Thermo Scientific Supplier No. 6850.1023
Sample rack, 54 pos, 12 mm OD vials, Thermo Scientific™ Vanquish™ Charger and Split Sampler Sample racks are for use in the Vanquish Split Sampler modules, and fit into the 9-shelf of the Charger module, which is an intelligent robotic that automates sample loading into the Thermo Scientific Vanquish UHPLC systems for extra-long unattended operation.