Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ Sample Rack for DK6/48 Digestion Unit
SDP

Catalog No. 11392141
Change view
Click to view available options
Positions:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
11-392-141 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-141 Supplier Velp™ Supplier No. A00000398
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sample rack for use with the DK6/48 Digestion Unit.

  • Conveniently hold samples for the DK6/48 Digestion Unit

Specifications

Autoclavable Not Autoclavable
Color Silver
Holds Tubes
Material Metal
Positions 6
Shape Vertical
Stackable No
Sterility Non-sterile
For Use With (Equipment) DK6/48 Digestion Unit
Product Type Vertical Tube Holder
Shelves None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.