Learn More
BURKLE INC Sampling scoop SteriPlast Bio-PE, sterile, 0.85 oz. (25 ml)
Catalog No. 503081280
Encompass_Preferred
Sampling scoop SteriPlast, sterile, made from eco-friendly green PE bio-plastic. This innovative Bio-PE material provides the same durability, chemical resistance, and food safety as conventional polyethylene, but is produced from renewable raw materials and is fully recyclable. Supplied sterile for hygienic, contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental testing. Capacity: 25 ml (0.85 oz).
Catalog No. 50-308-1280 Supplier BURKLE INC Supplier No. 53791001
Sampling scoop SteriPlast, sterile, made from eco-friendly green PE bio-plastic. This innovative Bio-PE material provides the same durability, chemical resistance, and food safety as conventional polyethylene, but is produced from renewable raw materials and is fully recyclable. Supplied sterile for hygienic, contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental testing. Capacity: 25 ml (0.85 oz).