Learn More
Thermo Scientific™ Sample Sensor for Thermo Scientific™ Heratherm™ Advanced Protocol Security Incubators
Catalog No. 13998201 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Sensor
Catalog No. Type
13-998-201 Sensor
1 options
Catalog No. 13-998-201 Supplier Thermo Scientific™ Supplier No. 50127768
Add to Cart
Add to Cart
Measures exact temperature
  • For connection to all Advanced Protocol Security incubators
  • Measures exact sample temperature
  • Temperature shown on display as plugged in
  • Cable length: 86.6 in.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Advanced Protocol Security Incubators
Type Sensor