Learn More
BURKLE INC Palette knife spatula, LxW 7.56 x 0.79 in (192 x 20 mm), PS, white
Catalog No. 503081416
Encompass_Preferred
The sample spatula can be used to apply, spread or smooth viscous media such as pastes or creams. The spatula easily reaches into the corners and curves of various containers and is suitable for scraping out bowls, removing residues or wiping off Bürkle's measuring and dosing spoons.
The dimensionally stable long edge of the plastic scraper is ideal for cutting viscous media. A small tip on the long edge can be used to tear packaging film, for example. The scraper is also suitable for stirring and mixing.
Catalog No. 50-308-1416 Supplier BURKLE INC Supplier No. 53780035
The sample spatula can be used to apply, spread or smooth viscous media such as pastes or creams. The spatula easily reaches into the corners and curves of various containers and is suitable for scraping out bowls, removing residues or wiping off Bürkle's measuring and dosing spoons.
The dimensionally stable long edge of the plastic scraper is ideal for cutting viscous media. A small tip on the long edge can be used to tear packaging film, for example. The scraper is also suitable for stirring and mixing.