Learn More
BURKLE INC Spatula SteriPlast, PS, sterile, 5.91 in. (150 mm), with cover
Catalog No. 503081259
Encompass_Preferred
The disposable spatula made of polystyrene is produced and assembled in our clean room of class 7 (10,000) and individually packaged for disposable use.
As a result of the long, ergonomically-shaped, stable handle and a sharp blade, it is possible to even penetrate directly into containers such as paper or plastic sacks.
Catalog No. 50-308-1259 Supplier BURKLE INC Supplier No. 53781010
The disposable spatula made of polystyrene is produced and assembled in our clean room of class 7 (10,000) and individually packaged for disposable use.
As a result of the long, ergonomically-shaped, stable handle and a sharp blade, it is possible to even penetrate directly into containers such as paper or plastic sacks.