Learn More
BURKLE INC Sample-spoon, V2A, 11.81 in. (300 mm), 0.30 oz. (9 ml), autoclavable
Catalog No. 5000026069
Encompass_Preferred
Made of high quality stainless steel 1.4301 (AISI 304), the Sample Spoon is the ideal solution for the reliable removal of small sample quantities in everyday laboratory work. It is ideal for precise work, even in hard-to-reach places.
The specially shaped spoon head ensures that almost every corner of the container can be reached - effectively preventing the sample from slipping. The ergonomically shaped handle with spatula blade fits securely in the hand for comfortable, controlled handling.
The narrow side of the spatula allows you to finely dispense the sample for precise portioning and analytical accuracy.
Catalog No. 50-000-26069 Supplier BURKLE INC Supplier No. 53200300
Made of high quality stainless steel 1.4301 (AISI 304), the Sample Spoon is the ideal solution for the reliable removal of small sample quantities in everyday laboratory work. It is ideal for precise work, even in hard-to-reach places.
The specially shaped spoon head ensures that almost every corner of the container can be reached - effectively preventing the sample from slipping. The ergonomically shaped handle with spatula blade fits securely in the hand for comfortable, controlled handling.
The narrow side of the spatula allows you to finely dispense the sample for precise portioning and analytical accuracy.