Learn More
Electron Microscopy Sciences Sample Storage Set, Sk-3
SDP
Catalog No. 5033085
Encompass_Preferred
Most economical, straight-sided, flat bottom, glass shell vial with high quality cork closures. Ideally suited for storing small parts, geological specimens, soil and many other types of samples. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Vial size 3 dr (10.4ml), 12 mm dia x 114 mm H, arrangement 5x20. Tray size 11 1/2" x 3" x 5" Vial size: 3 dr (10.4ml), 12mm dia x 114mm H, arrangement: 5x20, tray size: 11-1⁄2" x 3"
Catalog No. 50-330-85 Supplier Electron Microscopy Sciences Supplier No. 64269
Most economical, straight-sided, flat bottom, glass shell vial with high quality cork closures. Ideally suited for storing small parts, geological specimens, soil and many other types of samples. Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card. Alpha-numerical indexing system allows quick identification and location of stored items. Vial size 3 dr (10.4ml), 12 mm dia x 114 mm H, arrangement 5x20. Tray size 11 1/2" x 3" x 5" Vial size: 3 dr (10.4ml), 12mm dia x 114mm H, arrangement: 5x20, tray size: 11-1⁄2" x 3"