Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CoaguSense™ Sample Transfer Tubes with Plunger

Catalog No. 23654005
Change view
Click to view available options
Quantity:
54
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-654-005 54
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-654-005 Supplier CoaguSense™ Supplier No. 03P5254

missing translation for 'validMainframeMsg'

CoaguSense™ Sample Transfer Tubes with Plunger offer excellent blood borne pathogen control and less test strip wastage

  • Glass tubes with plastic clad
  • Includes plunger
  • 54 clad
  • Delivers the right volume of capillary blood to the Coag-Sense™ system

Specifications

Content And Storage Room Temperature
Format Transfer Tubes
Type Capillary
Includes Plunger
Material Glass, Plastic
Packaging Type 12 μL Tubes
Quantity 54
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.