Learn More
Harrick Plasma Pyrex Sample Tray for PDC-32G
SDP
Catalog No. NC1585286
Encompass_Preferred
The PDC-32T-P Quartz Sample Tray is 2.75"W x 6.5"L x 0.125” thick and provides a horizontal surface for loading small samples and for batch processing. Use with the PDC-32G Basic Plasma Cleaner.
Catalog No. NC1585286 Supplier Harrick Plasma Supplier No. PDC32TP
The PDC-32T-P Quartz Sample Tray is 2.75"W x 6.5"L x 0.125” thick and provides a horizontal surface for loading small samples and for batch processing. Use with the PDC-32G Basic Plasma Cleaner.