Learn More
Electron Microscopy Sciences Micro Sample Tube Rack with 20 Holes 7.7 mm each, 105 mm
SDP
Catalog No. 501899859
Encompass_Preferred
These round floating bubble racks can incubate 8 to 20 tubes in a beaker of water. The racks are loaded from the top and the tubes are easily removed by pressing the rack down on a bench until the short legs touch. Perfect for use in microbiology and immunology labs.
Molded from buoyant foam-filled TPXR (polymethylpentene) with tiny bubbles throughout. 10.8 mm diameter holes fit the 1.5 ml microtubes, and 7.7 mm holes fit the 0.5 ml tube. The racks are 1/4"(6.4 mm) thick with 3/4" (19 mm) long legs and a center rod handle. They are steam autoclavable at 250 degree F (121 degree C). Use in 1000ml beakers. 7.7mm holes fit the 0.5ml tube.
Hole: 7.7 mm
O.D.: 105 mm
Use: 1000 mL beaker
Catalog No. 50-189-9859 Supplier Electron Microscopy Sciences Supplier No. 7236422
Add to Cart
Add to Cart
These round floating bubble racks can incubate 8 to 20 tubes in a beaker of water. The racks are loaded from the top and the tubes are easily removed by pressing the rack down on a bench until the short legs touch. Perfect for use in microbiology and immunology labs.
Molded from buoyant foam-filled TPXR (polymethylpentene) with tiny bubbles throughout. 10.8 mm diameter holes fit the 1.5 ml microtubes, and 7.7 mm holes fit the 0.5 ml tube. The racks are 1/4"(6.4 mm) thick with 3/4" (19 mm) long legs and a center rod handle. They are steam autoclavable at 250 degree F (121 degree C). Use in 1000ml beakers. 7.7mm holes fit the 0.5ml tube.
Hole: 7.7 mm
O.D.: 105 mm
Use: 1000 mL beaker