Learn More
Chemglass Life Sciences Sample Tubes, 50mL, with 18'' C-Flex Inlet, C-Flex Dip Tube, Amber, Sterile, Bioprocess Sampling, Single Use
SDP
Catalog No. 5000083109
Encompass
The 50mL amber polypropylene centrifuge tube assembly utilizes a closed cap assembly with 18 of size 16 (1/8' x 1/4) C-Flex tubing inlet line with a pinch clamp and plugged.The assembly includes a 3 vent line with a PTFE filter.Assembly is certified sterile. The assembly can be used for sampling and transfer of biopharmaceutical products and reagents in closed systems.Sold individually wrapped in a bag of ten.
Catalog No. 50-000-83109 Supplier Chemglass Life Sciences Supplier No. CLS1407T312A
The 50mL amber polypropylene centrifuge tube assembly utilizes a closed cap assembly with 18 of size 16 (1/8' x 1/4) C-Flex tubing inlet line with a pinch clamp and plugged.The assembly includes a 3 vent line with a PTFE filter.Assembly is certified sterile. The assembly can be used for sampling and transfer of biopharmaceutical products and reagents in closed systems.Sold individually wrapped in a bag of ten.