Learn More
Chemglass Life Sciences Sample Tubes, 50mL, with 18'' C-Flex Inlet, C-Flex Dip Tube, Tygon Seal Tube, Gamma, Bioprocess Sampling, Single Use
SDP
Catalog No. 502741238
Encompass
The 50mL polypropylene centrifuge tube assembly utilizes a closed cap assembly with 3/4 of size 16 (1/8' x 1/4) C-Flex tubing inlet line which transitions to 2 of Size 14 (1/16 x ) Tygon and then transitions to 17 of size 16 (1/8' x 1/4) C-Flex tubing with a pinch clamp and plugged.The assembly includes a 3 vent line with a PTFE filter.Assembly is certified sterile.The assembly can be used for sampling and transfer of biopharmaceutical products and reagents in closed systems.Sold individually wrapped in a bag of ten.
Catalog No. 50-274-1238 Supplier Chemglass Life Sciences Supplier No. CLS1407T310
The 50mL polypropylene centrifuge tube assembly utilizes a closed cap assembly with 3/4 of size 16 (1/8' x 1/4) C-Flex tubing inlet line which transitions to 2 of Size 14 (1/16 x ) Tygon and then transitions to 17 of size 16 (1/8' x 1/4) C-Flex tubing with a pinch clamp and plugged.The assembly includes a 3 vent line with a PTFE filter.Assembly is certified sterile.The assembly can be used for sampling and transfer of biopharmaceutical products and reagents in closed systems.Sold individually wrapped in a bag of ten.