Learn More
Fritsch Milling And Sizing Inc Sample Vessel 25Ml for Sample Bottles 250 And 500Ml
SDP
Catalog No. 501534709
Encompass
Sample vessel 25 ml with lid for sample bottle 250 ml and 500 ml
For dividing smallest sample quantities, this sample vessel 25 ml can be inserted in the sample bottles 250 ml (article-number 27.1450.00) and sample bottles 500 ml (article-number 27.1460.00).
Catalog No. 50-153-4709 Supplier Fritsch Milling And Sizing Inc Supplier No. 83.3190.16
Sample vessel 25 ml with lid for sample bottle 250 ml and 500 ml
For dividing smallest sample quantities, this sample vessel 25 ml can be inserted in the sample bottles 250 ml (article-number 27.1450.00) and sample bottles 500 ml (article-number 27.1460.00).