Learn More
Hach Company Sample Vial, 120 mL with Thiosulfate and Custody Tie, 200/pk
Catalog No. 501996398 Shop All Hach Company Products
Encompass
120 mL sterile, hinged-cap vials meet all EPA requirements for microbiology sampling. Collection, incubation, and accurate reading can all be done in the vial without a transfer step. 100 mL raised fill line. Polypropylene.
Catalog No. 50-199-6398 Supplier Hach Company Supplier No. 2634001
120 mL sterile, hinged-cap vials meet all EPA requirements for microbiology sampling. Collection, incubation, and accurate reading can all be done in the vial without a transfer step. 100 mL raised fill line. Polypropylene.