Learn More
Hach Company Sample Vial, 120 mL without Thiosulfate, 200/pk
Supplier:  Hach Company 2634002
Encompass
120 mL sterile, hinged-cap vials meet all EPA requirements for microbiology sampling. Collection, incubation, and accurate reading can all be done in the vial without a transfer step. 100 mL raised fill line. Polypropylene.
Catalog No. 50-199-6399