Learn More
Metrohm USA SAMPLE VIAL 6.2419.000 6ML
SDP
Catalog No. NC0251440 Shop All Metrohm USA Products
Encompass
Sample vial 6 mL / 100 vials per pack. Suitable for the 6.1448.05X septum. Utilization for KF determinations with oven.
Catalog No. NC0251440 Supplier Metrohm USA Supplier No. 62419007
Sample vial 6 mL / 100 vials per pack. Suitable for the 6.1448.05X septum. Utilization for KF determinations with oven.