Learn More
Electron Microscopy Sciences Wheaton 320 Borosilicate Sample Vial, Amber, 4 ml, 15 x 48 mm
SDP
Catalog No. 501899005
Encompass_Preferred
Multi purpose glass sample vials specifically suited for research studies. These sample vials are economically priced and conveniently packaged in two shrink-wrapped packs of 100 vials. These vials come with lined caps and are packaged separately. The vials can be sterilized and stored at low temperatures. Wheaton 320 borosilicate glass protects light-sensitive samples.
Color: Amber
Volume: 4 ml
Size: 15 x 48 mm
Catalog No. 50-189-9005 Supplier Electron Microscopy Sciences Supplier No. 6099304
Multi purpose glass sample vials specifically suited for research studies. These sample vials are economically priced and conveniently packaged in two shrink-wrapped packs of 100 vials. These vials come with lined caps and are packaged separately. The vials can be sterilized and stored at low temperatures. Wheaton 320 borosilicate glass protects light-sensitive samples.
Color: Amber
Volume: 4 ml
Size: 15 x 48 mm