Learn More
Electron Microscopy Sciences Wheaton 33R Borosilicate Sample Vial, Clear, 4 ml, 15 x 48 mm
SDP
Catalog No. 501898998
Encompass_Preferred
Multi purpose glass sample vials specifically suited for research studies. These sample vials are economically priced and conveniently packaged in two shrink-wrapped packs of 100 vials. These vials come with lined caps and are packaged separately. The vials can be sterilized and stored at low temperatures. Wheaton-33R low extractable borosilicate glass protects against changes in pH.
Color: Clear
Volume: 4 ml
Size: 15 x 48 mm
Catalog No. 50-189-8998 Supplier Electron Microscopy Sciences Supplier No. 6099204
Multi purpose glass sample vials specifically suited for research studies. These sample vials are economically priced and conveniently packaged in two shrink-wrapped packs of 100 vials. These vials come with lined caps and are packaged separately. The vials can be sterilized and stored at low temperatures. Wheaton-33R low extractable borosilicate glass protects against changes in pH.
Color: Clear
Volume: 4 ml
Size: 15 x 48 mm