Learn More
Hach Company Sample Vial, Glass, 11 mm, 10 per Pack
Supplier:  Hach Company LZM293
Encompass
Glass sample vial designed for laboratory sample collection, storage, and analysis. Supplied in packs of 10 with an 11 mm opening, these vials are suitable for use with common colorimeters and spectrophotometers or for general laboratory sampling needs.
  • Glass construction for chemical compatibility and clear visibility.
  • 11 mm opening to fit standard instrument sample holders.
  • Sold in packs of 10 for routine sampling and replacement needs.
  • Suitable for sample collection, storage, and spectrophotometric analysis.
  • Disposable or reusable depending on laboratory cleaning protocols.
Catalog No. NC2544158