Learn More
Lonza Walkersville Sample Containers
Catalog No. 501131990
Encompass
UPDATE (Apr 2019):We have decided to discontinue the performance of a sterility test for our Depyrogenated Sample Containers.As part of the production process, all glassware is depyrogenated at extremely high temperatures well in excess of standard depyrogenation process conditions. Additionally, the caps are terminally sterilized by dry or moist heat. These conditions provide for ample assurance that the sample containers are depyrogenated making them suitable for use when performing the Bacterial Endotoxins Test (BET). Therefore, there is no added value or additional quality assurance in subsequently performing a sterility test after processing. As a result, the Certificate of Analyses will no longer indicate the results of a sterility test for these products.Benefits:Certified to contain less than 0.005 EU/mlApplications:Transportation or storage of sample for endotoxin analysis
Catalog No. 50-113-1990 Supplier Lonza Walkersville Supplier No. 80507U
UPDATE (Apr 2019):We have decided to discontinue the performance of a sterility test for our Depyrogenated Sample Containers.As part of the production process, all glassware is depyrogenated at extremely high temperatures well in excess of standard depyrogenation process conditions. Additionally, the caps are terminally sterilized by dry or moist heat. These conditions provide for ample assurance that the sample containers are depyrogenated making them suitable for use when performing the Bacterial Endotoxins Test (BET). Therefore, there is no added value or additional quality assurance in subsequently performing a sterility test after processing. As a result, the Certificate of Analyses will no longer indicate the results of a sterility test for these products.Benefits:Certified to contain less than 0.005 EU/mlApplications:Transportation or storage of sample for endotoxin analysis