Learn More
Chemglass Life Sciences Vial, Sample, Pre-Cleaned, 40mL, Clear, 28x95mm, GPI 24-400, with PTFE Lined Screw Cap
SDP
Catalog No. 50230665
Encompass_Preferred
Vial, Sample, Pre-Cleaned, 40mL, Clear, 28x95mm, GPI 24-400, with PTFE Lined Screw Cap. Clear and amber sample vials are pre-cleaned and certified for volatile organic analysis. Vials are fully assembled with green polypropylene closure and 0.125” PTFE silicone septa. Made from Type 1 borosilicate glass. Certificate of analysis is included in each case. Packed 72 pieces per case.
Catalog No. 50-230-665 Supplier Chemglass Life Sciences Supplier No. CG490702
Vial, Sample, Pre-Cleaned, 40mL, Clear, 28x95mm, GPI 24-400, with PTFE Lined Screw Cap. Clear and amber sample vials are pre-cleaned and certified for volatile organic analysis. Vials are fully assembled with green polypropylene closure and 0.125” PTFE silicone septa. Made from Type 1 borosilicate glass. Certificate of analysis is included in each case. Packed 72 pieces per case.