Learn More
SEPSOLUTIONS SAMPLE VIALS 100UL 100/PK
Catalog No. NC9626594
Encompass_Preferred
350 microliter TPX transparent sample vial with snap ring top and conical profile interior, total volume 350 microliter with usable volume of 225 microliter, compatible with 11mm snap cap/crimp cap, pack of 100.
Catalog No. NC9626594 Supplier SEPSOLUTIONS Supplier No. 503012
350 microliter TPX transparent sample vial with snap ring top and conical profile interior, total volume 350 microliter with usable volume of 225 microliter, compatible with 11mm snap cap/crimp cap, pack of 100.