Learn More
METTLER TOLEDO 3P INDUSTRIAL SAMPLE VIALS 20ML 148PC
Catalog No. NC9695491
Encompass
Sample vials 20 mL made of borosilicate glass. • Item Packaging: 148 per case• Material: Borosilicate Glass• Capacity: 20ml• Dimensions: -• Vial Type: Sample• Bottom Type: Flat• Color: Clear• Cap Type: No cap• Cap Size: NA• Unit: Case• Qty: 148
Catalog No. NC9695491 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 51322310
Sample vials 20 mL made of borosilicate glass. • Item Packaging: 148 per case• Material: Borosilicate Glass• Capacity: 20ml• Dimensions: -• Vial Type: Sample• Bottom Type: Flat• Color: Clear• Cap Type: No cap• Cap Size: NA• Unit: Case• Qty: 148