Learn More
Chemglass Life Sciences 30mL Vial, 28mm OD x 70mm Height, 24-400 Thread Size, with PTFE Lined Solid Caps
SDP
Catalog No. CG490005
Encompass_Preferred
30mL Scintillation Vial, 28mm OD x 70mm Height, 24-400 Thread Size. Sample vials packaged in convenient LAB-PACS of 144. Made from Type 1 Borosilicate glass. Full height partitions minimize breakage. Solid PTFE Lined Caps.
Catalog No. CG490005 Supplier Chemglass Life Sciences Supplier No. CG490005
30mL Scintillation Vial, 28mm OD x 70mm Height, 24-400 Thread Size. Sample vials packaged in convenient LAB-PACS of 144. Made from Type 1 Borosilicate glass. Full height partitions minimize breakage. Solid PTFE Lined Caps.