Learn More
Electron Microscopy Sciences Sample Vials in Mini-File, 2 ml
SDP
Catalog No. 5028445
Encompass_Preferred
Comes ten vials packed in a clean plastic thermoformed Mini-File and offers convenience in transporting, displaying and storing. The vials are made from Wheaton-33R a low extractable borosilicate glass. The vials are supplied with a white (styrene-butadiene) rubber lined cap and both the vials and the caps can be autoclaved. Size: 12 dia x 37mm hgt with cap on.
Catalog No. 50-284-45 Supplier Electron Microscopy Sciences Supplier No. 6099602
Comes ten vials packed in a clean plastic thermoformed Mini-File and offers convenience in transporting, displaying and storing. The vials are made from Wheaton-33R a low extractable borosilicate glass. The vials are supplied with a white (styrene-butadiene) rubber lined cap and both the vials and the caps can be autoclaved. Size: 12 dia x 37mm hgt with cap on.