Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEX™ SamplePrep DNAmax Isolation Kit

Catalog No. 2402PT
Change view
Click to view available options
Sample Type:
Animal Tissue
Soft Plant Tissue
Tough Plant Tissue
Buffer:
Lysis, Wash, Elution
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Buffer
2402PT Tough Plant Tissue Lysis, Wash, Elution
2401M Animal Tissue Lysis, Wash, Elution
2402P Soft Plant Tissue Lysis, Wash, Elution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 2402PT Supplier SPEX™ Supplier No. 2402PT

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spex™ Genomic DNA Isolation Kits are optimized to work with the Genomax, Geno/Grinder, and GenoLyte and allow for fast and seamless isolation of DNA from a wide variety of animal tissues and cells, including mammalian, poultry, fish, and soft and tough plant tissue.

  • The complete kit solution for DNA extraction of animal and plant tissue
    • Reinforced tubes designed for homogenization through rapid beat beating
    • Silica-based columns capture DNA and allow for its purification
    • Protocol and workflow optimized for Genomax, Geno/Grinder, and GenoLyte

Specifications

Buffer Lysis, Wash, Elution
Bead Type Stainless Steel
Sample Type Tough Plant Tissue
For Use With (Equipment) Genomax, Geno/Grinder, GenoLyte
For Use With (Application) DNA Extraction of Animal and Plant Tissue
Includes Lysis Buffer, Binding Buffer B1, Wash Buffer PW1, Wash Buffer PW2, Elution Buffer, Proteinase K Powder, Rnase A, and Pre-Loaded Tubes with One 6 mm 440 C Stainless Steel Ball
No. of Reactions 75
Product Type DNA Isolation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.