Learn More
Cole-Parmer Nasco Sampling B01014WA Carrying Rack for 20, 18 oz Whirl-Pak Bags or 2.5" Diameter Bottles
Catalog No. NC9916364 Shop All Cole-Parmer Products
Encompass_Preferred
Sample Bottle Carrying Rack, Dimensions 13-1/4 in. L x 10- 3/4 in. W x 4 in. D Use rack with sample bottles 06043-01, 06049-01, 06050-01, 06020-64 on this page, or any bottle with maximum OD of 21 1/42 in. . Autoclavable. Rack: double grid
Catalog No. NC9916364 Supplier Cole-Parmer Supplier No. 6620300
Sample Bottle Carrying Rack, Dimensions 13-1/4 in. L x 10- 3/4 in. W x 4 in. D Use rack with sample bottles 06043-01, 06049-01, 06050-01, 06020-64 on this page, or any bottle with maximum OD of 21 1/42 in. . Autoclavable. Rack: double grid