Learn More
Hach Company Set of 24 1 L Polyethylene Bottles with Caps - Replacement bottles for portable samplers
Catalog No. NC2225377 Shop All Hach Company Products
Encompass
Set of 24 one-liter polyethylene sampling bottles with screw caps, sold as a replacement bottle set for compatible portable and refrigerated samplers. Each bottle provides 1 L of capacity and is supplied with a cap to support routine sample collection and storage.
  • Made from polyethylene.
  • 1 L capacity per bottle.
  • Includes 24 bottles with screw caps.
  • Sold as a replacement bottle set for compatible portable and refrigerated samplers.
  • Intended for sample collection, transport, and short-term storage.
This item is not returnable. View return policy
Catalog No. NC2225377 Supplier Hach Company Supplier No. 737
Set of 24 one-liter polyethylene sampling bottles with screw caps, sold as a replacement bottle set for compatible portable and refrigerated samplers. Each bottle provides 1 L of capacity and is supplied with a cap to support routine sample collection and storage.
  • Made from polyethylene.
  • 1 L capacity per bottle.
  • Includes 24 bottles with screw caps.
  • Sold as a replacement bottle set for compatible portable and refrigerated samplers.
  • Intended for sample collection, transport, and short-term storage.